Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib hydrochloride dihydrate | 99.9% | 602.01 g/mol | C28H27ClF3N7O3 | 50 MG
SDP

Catalog No. 5000249255
Encompass_Preferred

Nilotinib hydrochloride dihydrate is a small-molecule Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity, supplied as a solid research reagent for preclinical studies including chronic myelogenous leukemia models. The product includes analytical characterization and handling guidance for laboratory use.

  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; ultrasonic aid recommended.
  • Solid form with molecular weight 602.01 g/mol.
  • Storage: room temperature guidance, 4°C sealed, solvent stability at -80°C/-20°C.
  • Suitable for in vitro and in vivo kinase inhibition studies.

Catalog No. 50-002-49255 Supplier Medchemexpress LLC Supplier No. HY10159C50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib hydrochloride dihydrate is a small-molecule Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity, supplied as a solid research reagent for preclinical studies including chronic myelogenous leukemia models. The product includes analytical characterization and handling guidance for laboratory use.

  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; ultrasonic aid recommended.
  • Solid form with molecular weight 602.01 g/mol.
  • Storage: room temperature guidance, 4°C sealed, solvent stability at -80°C/-20°C.
  • Suitable for in vitro and in vivo kinase inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.