Learn More
Medchemexpress LLC HY-10159A 100mg , Nilotinib (monohydrochloride monohydrate) CAS:923288-90-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10159A100MG
Encompass_Preferred
HY-10159A 100mg Nilotinib (monohydrochloride monohydrate) CAS: 923288-90-8 Nilotinib monohydrochloride monohydrate is a second generation tyrosine kinase inhibitor (TKI), is significantly potent against BCR-ABL, and is active against many BCR-ABL mutants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9425