Learn More
Apexbio Technology LLC Nilotinib monohydrochloride monohydrate 923288-90-8 250mg
SDP
Catalog No. 5000093419
Encompass_Preferred
Nilotinib monohydrochloride monohydrate (CAS 923288-90-8) is a selective small molecule inhibitor targeting the BCR-ABL tyrosine kinase By binding to the ATP-binding site of BCR-ABL nilotinib effectively suppresses kinase activity impeding downstream signaling involved in cell proliferation It demonstrates potent inhibitory activity with an IC50 below 30 nM Nilotinib is widely utilized in biomedical research to investigate BCR-ABL-mediated cellular mechanisms and to model therapeutic interventions in chronic myelogenous leukemia and related malignancies
Catalog No. 50-000-93419 Supplier Apexbio Technology LLC Supplier No. A3660250
Nilotinib monohydrochloride monohydrate (CAS 923288-90-8) is a selective small molecule inhibitor targeting the BCR-ABL tyrosine kinase By binding to the ATP-binding site of BCR-ABL nilotinib effectively suppresses kinase activity impeding downstream signaling involved in cell proliferation It demonstrates potent inhibitory activity with an IC50 below 30 nM Nilotinib is widely utilized in biomedical research to investigate BCR-ABL-mediated cellular mechanisms and to model therapeutic interventions in chronic myelogenous leukemia and related malignancies