Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib monohydrochloride monohydrate | 923288-90-8 | MFCD18251359 | >98.0% | 583.99 g/mol | C28H25ClF3N7O2 | 200 MG
SDP

Catalog No. 5000247153
Encompass_Preferred

Nilotinib monohydrochloride monohydrate is the monohydrochloride monohydrate salt of nilotinib, a second-generation Bcr-Abl tyrosine kinase inhibitor used in research to study Bcr-Abl-driven signaling and oncology applications. Provided as a high-purity research compound, it is suitable for biochemical and cellular assays investigating kinase inhibition and resistance mutations.

  • High purity suitable for research applications.
  • Soluble in DMSO; limited solubility in water.
  • Active against Bcr-Abl and many resistance-associated mutants.
  • Molecular weight 583.99 g/mol for formulation and dosing calculations.
  • Supplied as a stable solid salt for ease of handling and storage.

Catalog No. 50-002-47153 Supplier Medchemexpress LLC Supplier No. HY10159A200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib monohydrochloride monohydrate is the monohydrochloride monohydrate salt of nilotinib, a second-generation Bcr-Abl tyrosine kinase inhibitor used in research to study Bcr-Abl-driven signaling and oncology applications. Provided as a high-purity research compound, it is suitable for biochemical and cellular assays investigating kinase inhibition and resistance mutations.

  • High purity suitable for research applications.
  • Soluble in DMSO; limited solubility in water.
  • Active against Bcr-Abl and many resistance-associated mutants.
  • Molecular weight 583.99 g/mol for formulation and dosing calculations.
  • Supplied as a stable solid salt for ease of handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.