Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib monohydrochloride monohydrate | 923288-90-8 | MFCD18251359 | 99.9% | 583.99 g·mol⁻¹ | C28H25ClF3N7O2 | 1 ML
SDP

Catalog No. 5000249110
Encompass_Preferred

Nilotinib monohydrochloride monohydrate is the hydrochloride monohydrate salt of nilotinib, a second-generation BCR-ABL tyrosine kinase inhibitor used in research to study BCR-ABL signaling, drug resistance, and mutant kinases. It is provided in concentrated stock-ready formats for biochemical, cell-based, and in vivo studies.

  • Second-generation BCR-ABL tyrosine kinase inhibitor with activity against common resistant mutants.
  • High chemical purity (~99.9% by LCMS) suitable for analytical and biological studies.
  • Hydrochloride monohydrate salt form for improved solubility and handling.
  • Molecular weight 583.99 g·mol⁻¹; formula C28H25ClF3N7O2.
  • Soluble in DMSO (≈57 mM) and in acidified aqueous media; supplier provides stock preparation guidance.
  • Intended for in vitro biochemical and cell-based assays and for in vivo formulation with appropriate vehicles.

Catalog No. 50-002-49110 Supplier Medchemexpress LLC Supplier No. HY10159A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib monohydrochloride monohydrate is the hydrochloride monohydrate salt of nilotinib, a second-generation BCR-ABL tyrosine kinase inhibitor used in research to study BCR-ABL signaling, drug resistance, and mutant kinases. It is provided in concentrated stock-ready formats for biochemical, cell-based, and in vivo studies.

  • Second-generation BCR-ABL tyrosine kinase inhibitor with activity against common resistant mutants.
  • High chemical purity (~99.9% by LCMS) suitable for analytical and biological studies.
  • Hydrochloride monohydrate salt form for improved solubility and handling.
  • Molecular weight 583.99 g·mol⁻¹; formula C28H25ClF3N7O2.
  • Soluble in DMSO (≈57 mM) and in acidified aqueous media; supplier provides stock preparation guidance.
  • Intended for in vitro biochemical and cell-based assays and for in vivo formulation with appropriate vehicles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.