Learn More
Apexbio Technology LLC Nilotinib(AMN-107) 641571-10-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061652
Encompass_Preferred
Nilotinib (AMN-107 CAS 641571-10-0) is an orally bioavailable selective inhibitor targeting the BCR-ABL tyrosine kinase Derived structurally from imatinib nilotinib inhibits both wild-type (WT p210) and mutant forms (E281K E292K F317L M351T F486S) of BCR-ABL reducing their autophosphorylation activity with IC50 values ranging from 20 to 42 nM It also exhibits substantial inhibitory activity against activated KIT mutants (e g V560del K642E) and various KIT double mutations and effectively suppresses PDGFR and PDGFR kinases Nilotinib is utilized extensively in research related to chronic myeloid leukemia and gastrointestinal stromal tumors involving kinase-driven pathologies
Catalog No. 50-000-61652 Supplier Apexbio Technology LLC Supplier No. A82325.1
Add to Cart
Add to Cart
Nilotinib (AMN-107 CAS 641571-10-0) is an orally bioavailable selective inhibitor targeting the BCR-ABL tyrosine kinase Derived structurally from imatinib nilotinib inhibits both wild-type (WT p210) and mutant forms (E281K E292K F317L M351T F486S) of BCR-ABL reducing their autophosphorylation activity with IC50 values ranging from 20 to 42 nM It also exhibits substantial inhibitory activity against activated KIT mutants (e g V560del K642E) and various KIT double mutations and effectively suppresses PDGFR and PDGFR kinases Nilotinib is utilized extensively in research related to chronic myeloid leukemia and gastrointestinal stromal tumors involving kinase-driven pathologies