Learn More
Enzo Life Sciences Niltubacin (500µg)
SDP
Supplier:  Enzo Life Sciences BMLGR3630500
Encompass_Preferred
Negative control for Tubacin (tubulin acetylation inducer), a highly potent and selective, reversible, cell-permeable inhibitor of HDAC6. Concentration of niltubacin in cell culture experiments typically ranges from 2-20µM. Alternative name: 7-(4-((2R,4R,6S)-4-((4,5-dipenyloxazol-2-ylthio)methyl)-6-(4-(hydroxymethyl)phenyl)-1,3-dioxan-2-yl)phenylcarbamoyl)heptanoic acid. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (6 mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-1015