Learn More
Medchemexpress LLC Nilutamide | 63612-50-0 | 99.7% | 317.22 | 200 MG
SDP
Catalog No. 5000348705
Encompass_Preferred
Nilutamide is an orally active nonsteroidal androgen receptor antagonist. It has affinity for androgen receptors but not for progestogen, estrogen, or glucocorticoid receptors. It can be used for research on prostate cancer and also exhibits antischistosomal properties. It inhibits hexobarbital hydroxylase, benzphetamine N-demethylase, benzo(a)pyrene hydroxylase, and 7-ethoxycoumarin O-deethylase activities in human liver microsomes.
  • Orally active nonsteroidal androgen receptor antagonist
  • Used for research on prostate cancer
  • Exhibits antischistosomal properties
  • Inhibits hexobarbital hydroxylase and other enzyme activities
Catalog No. 50-003-48705 Supplier Medchemexpress LLC Supplier No. HY13702200MG
Nilutamide is an orally active nonsteroidal androgen receptor antagonist. It has affinity for androgen receptors but not for progestogen, estrogen, or glucocorticoid receptors. It can be used for research on prostate cancer and also exhibits antischistosomal properties. It inhibits hexobarbital hydroxylase, benzphetamine N-demethylase, benzo(a)pyrene hydroxylase, and 7-ethoxycoumarin O-deethylase activities in human liver microsomes.
  • Orally active nonsteroidal androgen receptor antagonist
  • Used for research on prostate cancer
  • Exhibits antischistosomal properties
  • Inhibits hexobarbital hydroxylase and other enzyme activities