Learn More
Medchemexpress LLC Nilutamide-d6 | 1189477-66-4 | 99.8% | 323.26 | 1 MG
SDP
Catalog No. 5000350716
Encompass_Preferred
Nilutamide-d6 (Nilandron-d6) is a deuterium labeled Nilutamide. It is a non-steroidal anti-androgen agent proposed in the research of metastatic prostatic carcinoma. Deuterium labeling incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily as tracers for quantitation during drug development. This deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-50716 Supplier Medchemexpress LLC Supplier No. HY13702S1MG
Nilutamide-d6 (Nilandron-d6) is a deuterium labeled Nilutamide. It is a non-steroidal anti-androgen agent proposed in the research of metastatic prostatic carcinoma. Deuterium labeling incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily as tracers for quantitation during drug development. This deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS