Learn More
Medchemexpress LLC Nilutamide-d6 | 1189477-66-4 | 99.8% | C12H4D6F3N3O4 | 10MG
SDP
Catalog No. 5000259097
Encompass_Preferred
Nilutamide-d6 is the deuterium-labeled analogue of nilutamide supplied as a solid research reagent and analytical internal standard for use in ADME, mass spectrometry, and pharmacokinetic studies. It is intended for research use only.
  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • High chemical purity for reliable analytical results.
  • Suitable for ADME and pharmacokinetic studies.
  • Solid powder form enables straightforward sample preparation.
  • Stable when stored sealed at 4°C, protected from moisture and light.
Catalog No. 50-002-59097 Supplier Medchemexpress LLC Supplier No. HY13702S10MG
Nilutamide-d6 is the deuterium-labeled analogue of nilutamide supplied as a solid research reagent and analytical internal standard for use in ADME, mass spectrometry, and pharmacokinetic studies. It is intended for research use only.
  • Deuterium-labeled internal standard for mass spectrometry quantitation.
  • High chemical purity for reliable analytical results.
  • Suitable for ADME and pharmacokinetic studies.
  • Solid powder form enables straightforward sample preparation.
  • Stable when stored sealed at 4°C, protected from moisture and light.