Learn More
Cayman Chemical Nilvadipine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2928067-26-7, 5 mg
SDP
Catalog No. 502949739
Encompass_Preferred
An internal standard for the quantification of nilvadipine by GC- or LC-MS
Catalog No. 50-294-9739 Supplier Cayman Chemical Supplier No. 337005 MG
An internal standard for the quantification of nilvadipine by GC- or LC-MS