Learn More
Medchemexpress LLC Nimbin | 5945-86-8 | 1 MG
SDP
Catalog No. 5000417760
Encompass_Preferred
Nimbin is an orally active intermediate limonoid found in Azadirachta. It prevents tau aggregation, increases cell viability, and effectively inhibits the envelope protein of the dengue virus. It shows promise for research in neurodegenerative diseases and viral infections.
  • Prevents tau aggregation
  • Increases cell viability
  • Effectively inhibits the envelope protein of the dengue virus
  • Possesses anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties
  • Promising for research in neurodegenerative diseases and viral infections
Catalog No. 50-004-17760 Supplier Medchemexpress LLC Supplier No. HYN31871MG
Nimbin is an orally active intermediate limonoid found in Azadirachta. It prevents tau aggregation, increases cell viability, and effectively inhibits the envelope protein of the dengue virus. It shows promise for research in neurodegenerative diseases and viral infections.
  • Prevents tau aggregation
  • Increases cell viability
  • Effectively inhibits the envelope protein of the dengue virus
  • Possesses anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties
  • Promising for research in neurodegenerative diseases and viral infections