Learn More
Medchemexpress LLC Nimbin | 5945-86-8 | 5 MG
SDP
Catalog No. 5000418085
Encompass_Preferred
Nimbin is an orally active intermediate limonoid found in Azadirachta. It prevents tau aggregation, increases cell viability, and effectively inhibits the envelope protein of the dengue virus. It possesses anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties, showing promise for research into neurodegenerative diseases and viral infections.
  • Orally active intermediate limonoid
  • Prevents tau aggregation and increases cell viability
  • Effective inhibitor of the dengue virus envelope protein
  • Anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties
  • Promising for research of neurodegenerative diseases and viral infections
Catalog No. 50-004-18085 Supplier Medchemexpress LLC Supplier No. HYN31875MG
Nimbin is an orally active intermediate limonoid found in Azadirachta. It prevents tau aggregation, increases cell viability, and effectively inhibits the envelope protein of the dengue virus. It possesses anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties, showing promise for research into neurodegenerative diseases and viral infections.
  • Orally active intermediate limonoid
  • Prevents tau aggregation and increases cell viability
  • Effective inhibitor of the dengue virus envelope protein
  • Anti-inflammatory, antipyretic, antifungal, antihistamine, antiseptic, antioxidant, anti-cancer, and anti-viral properties
  • Promising for research of neurodegenerative diseases and viral infections