Learn More
Medchemexpress LLC HY-116035 5mg , Nimbolide CAS:25990-37-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1160355MG
Encompass_Preferred
HY-116035 5mg Nimbolide CAS: 25990-37-8 Nimbolide is a triterpene derived from the leaves and flowers of neem (Azadirachta indica L). Nimbolide induces apoptosis through inactivation of NF-κB. Nimbolide inhibits CDK4/CDK6 kinase activity. Nimbolide suppresses the NF-κB, Wnt, PI3K-Akt, MAPK and JAK-STAT signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1306