Learn More
Medchemexpress LLC HY-B0363 100mg Medchemexpress, Nimesulide CAS:51803-78-2 Purity:>98%
SDP
Catalog No. 502082491
Encompass_Preferred
Medchemexpress, HY-B0363 100mg Nimesulide CAS:51803-78-2 Nimesulide is a selective COX-2 inhibitor, with IC50s of 70 nM-70 μM in a time-dependent manner, but it shows no effect on COX-1 (IC50 >100 μM). Nimesulide has potent anti-inflammatory, analgesic and antipyretic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2491 Supplier Medchemexpress LLC Supplier No. HYB0363100MG
Medchemexpress, HY-B0363 100mg Nimesulide CAS:51803-78-2 Nimesulide is a selective COX-2 inhibitor, with IC50s of 70 nM-70 μM in a time-dependent manner, but it shows no effect on COX-1 (IC50 >100 μM). Nimesulide has potent anti-inflammatory, analgesic and antipyretic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.