Learn More
Apexbio Technology LLC Nimesulide 51803-78-2 100mg
SDP
Supplier:  Apexbio Technology LLC B1452100
Encompass_Preferred
Nimesulide (CAS 51803-78-2) is a non-steroidal anti-inflammatory drug (NSAID) characterized by preferential inhibition of cyclooxygenase-2 (COX-2) activity By selectively targeting COX-2 nimesulide reduces the synthesis of pro-inflammatory prostaglandins thereby exerting anti-inflammatory analgesic and antipyretic effects Due to its COX-2 selectivity nimesulide has been investigated in various models of inflammation pain and fever This compound is commonly used in preclinical and pharmacological studies to explore COX-2 mediated pathways and evaluate potential therapeutic strategies for inflammatory disorders
Catalog No. 50-224-0624