Learn More
Enzo Life Sciences Nimodipine (1g). CAS: 66085-59-4
SDP
Supplier:  Enzo Life Sciences ALX550277G001
Encompass_Preferred
Ca2+ channel antagonist. Highly potent at L-type channels. Block some low-voltage activated Ca2+ channels. Enhances memory in old or brain-damaged animals. Ameliorates experimental diabetic neuropathy in streptozotocin-induced diabetic rats. Vasodilator. Alternative name: 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl-1-methylethyl ester. Purity: ≥99% (Assay). Appearance: Yellowish crystalline powder. Solubility: Soluble in DMSO (25mg/ml) or methanol. Long Term Storage: Ambient.
Catalog No. 50-200-9521