Learn More
Medchemexpress LLC Nimodipine | 66085-59-4 | 99.9% | 1 ML
SDP
Catalog No. 5000406889
Encompass_Preferred
Nimodipine is an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist. It is primarily used for the research of cerebrovascular disorders and is intended for research use only.
  • Orally active and well-tolerated
  • Light-sensitive dihydropyridine calcium antagonist
  • Used in research for cerebrovascular disorders
  • Available as a light yellow to yellow solid
  • For research use only
Catalog No. 50-004-06889 Supplier Medchemexpress LLC Supplier No. HYB026510MM/1ML
Nimodipine is an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist. It is primarily used for the research of cerebrovascular disorders and is intended for research use only.
  • Orally active and well-tolerated
  • Light-sensitive dihydropyridine calcium antagonist
  • Used in research for cerebrovascular disorders
  • Available as a light yellow to yellow solid
  • For research use only