Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nimodipine | 66085-59-4 | MFCD00153848 | 99.9% | 500 MG
SDP

Catalog No. 5000406986
Encompass_Preferred

Nimodipine is an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist used for the research of cerebrovascular disorders.

  • Orally active and well-tolerated dihydropyridine calcium antagonist.
  • Light-sensitive compound.
  • Molecular formula: C21H26N2O7.
  • Molecular weight: 418.44.
  • Appearance: Solid, light yellow to yellow.
  • Shipping at room temperature.
  • Storage at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO: 100 mg/mL.

Catalog No. 50-004-06986 Supplier Medchemexpress LLC Supplier No. HYB0265500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nimodipine is an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist used for the research of cerebrovascular disorders.

  • Orally active and well-tolerated dihydropyridine calcium antagonist.
  • Light-sensitive compound.
  • Molecular formula: C21H26N2O7.
  • Molecular weight: 418.44.
  • Appearance: Solid, light yellow to yellow.
  • Shipping at room temperature.
  • Storage at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO: 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.