Learn More
Medchemexpress LLC Nimodipine-d7 | 1246815-36-0 | 99.4% | 1 MG
SDP
Catalog No. 5000405887
Encompass_Preferred
Nimodipine-d7 is the deuterium labeled Nimodipine, an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist primarily used for cerebrovascular disorder research. This compound serves as a valuable tool in drug development and quantitative analysis due to its deuterium labeling.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05887 Supplier Medchemexpress LLC Supplier No. HYB0265S1MG
Nimodipine-d7 is the deuterium labeled Nimodipine, an orally active, well-tolerated, and light-sensitive dihydropyridine calcium antagonist primarily used for cerebrovascular disorder research. This compound serves as a valuable tool in drug development and quantitative analysis due to its deuterium labeling.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs