Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nimodipine, EP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604827 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-827 1 g Amber Glass Bottle
18-604-828 5 g Amber Glass Bottle
18-604-829 25 g Amber Glass Bottle
18-604-830 100 g Amber Glass Bottle
18-604-831 1 kg Amber Glass Bottle
18-604-832 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-827 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N12841GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

N1284, 66085-59-4, C21H26N2O7

Spectrum™ Chemical Nimodipine, EP is a calcium channel blocker used to decrease problems due to a certain type of bleeding in the brain. All Spectrum Chemical EP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 66085-59-4
Molecular Formula C21H26N2O7
Molecular Weight (g/mol) 418.45
MDL Number MFCD00153848
InChI Key UIAGMCDKSXEBJQ-UHFFFAOYNA-N
IUPAC Name 3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C

Specifications

CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 g
Optical Rotation -0.10° to +0.10°
Percent Purity 98.5 to 101.5%
Grade EP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Nimodipine
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.