Learn More
Medchemexpress LLC HY-13703A 5mg Medchemexpress, Nimustine (hydrochloride) CAS:55661-38-6 Purity:>98%
SDP
Catalog No. 501932677
Encompass_Preferred
Medchemexpress, HY-13703A 5mg Nimustine (hydrochloride) CAS:55661-38-6 Nimustine hydrochloride (ACNU) is a DNA cross-linking and DNA alkylating agent, which induces DNA replication blocking lesions and DNA double-strand breaks and inhibits DNA synthesis , commonly used in chemotherapy for glioblastomas [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2677 Supplier Medchemexpress LLC Supplier No. HY13703A5MG
Medchemexpress, HY-13703A 5mg Nimustine (hydrochloride) CAS:55661-38-6 Nimustine hydrochloride (ACNU) is a DNA cross-linking and DNA alkylating agent, which induces DNA replication blocking lesions and DNA double-strand breaks and inhibits DNA synthesis , commonly used in chemotherapy for glioblastomas [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.