Learn More
Selleck Chemical LLC Nimustine Hydrochloride S5293-25mg
SDP
Catalog No. 503111925
Encompass_Preferred
Nimustine Hydrochloride (Nidran) is the hydrochloride salt of nimustine a nitrosourea with antineoplastic activity that alkylates and crosslinks DNA thereby causing DNA fragmentation inhibition of protein synthesis and cell death
Catalog No. 50-311-1925 Supplier Selleck Chemical LLC Supplier No. S529325MG
Nimustine Hydrochloride (Nidran) is the hydrochloride salt of nimustine a nitrosourea with antineoplastic activity that alkylates and crosslinks DNA thereby causing DNA fragmentation inhibition of protein synthesis and cell death