Learn More
Medchemexpress LLC Nimustine (hydrochloride) | 55661-38-6 | 98.2% | 309.15 | 25 MG
SDP
Catalog No. 5000351958
Encompass_Preferred
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. It functions as an alkylating agent, inducing DNA double-strand breaks and inter-strand crosslinks. This action activates the DNA damage response signaling pathway and the p38 MAPK/JNK signaling pathway, demonstrating antitumor activity.
  • Acts as an alkylating agent
  • Induces DNA double-strand breaks and inter-strand crosslinks
  • Activates DNA damage response signaling pathway
  • Activates p38 MAPK/JNK signaling pathway
  • Exhibits antitumor activity
  • Induces apoptosis in LN-229 cells
  • Inhibits tumor growth in C3H/HeN mouse models
Catalog No. 50-003-51958 Supplier Medchemexpress LLC Supplier No. 25MGHY13703A
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. It functions as an alkylating agent, inducing DNA double-strand breaks and inter-strand crosslinks. This action activates the DNA damage response signaling pathway and the p38 MAPK/JNK signaling pathway, demonstrating antitumor activity.
  • Acts as an alkylating agent
  • Induces DNA double-strand breaks and inter-strand crosslinks
  • Activates DNA damage response signaling pathway
  • Activates p38 MAPK/JNK signaling pathway
  • Exhibits antitumor activity
  • Induces apoptosis in LN-229 cells
  • Inhibits tumor growth in C3H/HeN mouse models