Learn More
Medchemexpress LLC Nimustine hydrochloride | 55661-38-6 | MFCD01676942 | 98.2% | C9H14Cl2N6O2 | 10MG
SDP
Catalog No. 5000259929
Encompass_Preferred
Nimustine hydrochloride is the hydrochloride salt of nimustine, an alkylating antitumor agent used in research to induce DNA damage and study DNA-damage response pathways. It produces DNA double-strand breaks and inter-strand crosslinks and activates signaling cascades involved in the DNA damage response.
Catalog No. 50-002-59929 Supplier Medchemexpress LLC Supplier No. HY13703A10MG
Nimustine hydrochloride is the hydrochloride salt of nimustine, an alkylating antitumor agent used in research to induce DNA damage and study DNA-damage response pathways. It produces DNA double-strand breaks and inter-strand crosslinks and activates signaling cascades involved in the DNA damage response.