Learn More
Medchemexpress LLC Nimustine hydrochloride (ACNU) | 55661-38-6 | 98.2% | 309.15 | 50 MG
SDP
Catalog No. 5000348718
Encompass_Preferred
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. This alkylating agent induces DNA double-strand breaks (DSBs) and inter-strand crosslinks (ICLs), activating the DNA damage response (DDR) signaling pathway. It also activates the p38 MAPK/JNK signaling pathway and demonstrates antitumor activity.
  • Exhibits cytotoxicity against human U87MG cells with an IC50 of 0.72 μM.
  • Activates caspase and AP-1, upregulates bim expression, and promotes c-Jun phosphorylation in LN-229 cells.
  • Induces apoptosis and increases expressions of cleaved bim and caspase-3/8/9 in LN-229 cells.
  • Inhibits tumor growth in C3H/HeN mouse models when combined with radiotherapy.
Catalog No. 50-003-48718 Supplier Medchemexpress LLC Supplier No. HY13703A50MG
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. This alkylating agent induces DNA double-strand breaks (DSBs) and inter-strand crosslinks (ICLs), activating the DNA damage response (DDR) signaling pathway. It also activates the p38 MAPK/JNK signaling pathway and demonstrates antitumor activity.
  • Exhibits cytotoxicity against human U87MG cells with an IC50 of 0.72 μM.
  • Activates caspase and AP-1, upregulates bim expression, and promotes c-Jun phosphorylation in LN-229 cells.
  • Induces apoptosis and increases expressions of cleaved bim and caspase-3/8/9 in LN-229 cells.
  • Inhibits tumor growth in C3H/HeN mouse models when combined with radiotherapy.