Learn More
Medchemexpress LLC Nimustine hydrochloride (ACNU) | 55661-38-6 | MFCD01676942 | 98.2% | 309.15 | 100 MG
SDP
Catalog No. 5000348707
Encompass_Preferred
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. It acts as an alkylating agent, inducing DNA double-strand breaks (DSBs) and inter-strand crosslinks (ICLs), which activates the DNA damage response (DDR) signaling pathway. Nimustine hydrochloride also activates the p38 MAPK/JNK signaling pathway and demonstrates antitumor activity.
  • Hydrochloride salt form of Nimustine
  • Alkylating agent
  • Induces DNA double-strand breaks and inter-strand crosslinks
  • Activates DNA damage response signaling pathway
  • Activates p38 MAPK/JNK signaling pathway
  • Exhibits antitumor activity
Catalog No. 50-003-48707 Supplier Medchemexpress LLC Supplier No. HY13703A100MG
Nimustine hydrochloride (ACNU) is the hydrochloride salt form of Nimustine. It acts as an alkylating agent, inducing DNA double-strand breaks (DSBs) and inter-strand crosslinks (ICLs), which activates the DNA damage response (DDR) signaling pathway. Nimustine hydrochloride also activates the p38 MAPK/JNK signaling pathway and demonstrates antitumor activity.
  • Hydrochloride salt form of Nimustine
  • Alkylating agent
  • Induces DNA double-strand breaks and inter-strand crosslinks
  • Activates DNA damage response signaling pathway
  • Activates p38 MAPK/JNK signaling pathway
  • Exhibits antitumor activity