Learn More
Cayman Chemical Nimustine (hydrochloride), Purity: ≥98%, CAS: 55661-38-6, 250 mg
SDP
Catalog No. 502991730
Encompass_Preferred
A pyrimidine analog and nitrosourea alkylating agent; inhibits the growth of U-251MG and EA285 glioma cells at 10 μg/ml; increases the median survival time in a Walker 256 carcinoma rat model of metastatic brain tumors at 30 mg/kg
Catalog No. 50-299-1730 Supplier Cayman Chemical Supplier No. 31464250 MG
A pyrimidine analog and nitrosourea alkylating agent; inhibits the growth of U-251MG and EA285 glioma cells at 10 μg/ml; increases the median survival time in a Walker 256 carcinoma rat model of metastatic brain tumors at 30 mg/kg