Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ NineFocus™ Analog Measurement Module (AMM) Conductivity

Catalog No. 01915887 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Conductivity Measurement Module
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-915-887 Conductivity Measurement Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-915-887 Supplier Mettler Toledo™ Supplier No. 31052300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for the use with NineFocus meters. It connects with InLab™ analog sensors to measure conductivity.

  • Backward compatible: Maximum flexibility for any application due to full compatibility with analog sensors (e.g., ISM) using an analog module
  • Designed to evolve: Expand the meter analysis productivity by adding additional modules. Promote sustainability Our high-quality modules are made from 100% recycled plastic, helping reduce the ecological footprint
  • Promote sustainability: Our high-quality modules are made from 100% recycled plastic, helping reduce the ecological footprint
TRUSTED_SUSTAINABILITY

Specifications

Calibration Automatic
Type Conductivity Measurement Module
Dimensions (L x W x H) 12 x 7 x 3.5 cm
IP Rating IP52
Length (Metric) 12 cm
Weight (Metric) 0.1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.