Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ NineFocus™ Cable Terminal 0.3m

Catalog No. 01915865 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Calibration:
Automatic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration
01-915-865 Automatic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-915-865 Supplier Mettler Toledo™ Supplier No. 31052304
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 0.3-meter cable provides a compact connection between the NineFocus meter and the control terminal, ideal for securely mounting the terminal in front of the meter.

  • Flexible footprint on bench: The cable connects to the detachable control terminal, allowing it to be placed flexibly thus increasing usability and convenience
TRUSTED_SUSTAINABILITY

Specifications

Calibration Automatic
Type Cable
IP Rating None
Length (Metric) 30 cm
Recommended Accessories Meter Accessories
Weight (Metric) 120 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.