Learn More
Mettler Toledo NineFocus Control Terminal
Catalog No. 01915890 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
NineFocus pH Meter
Catalog No. For Use With (Equipment)
01-915-890 NineFocus pH Meter
1 options
Catalog No. 01-915-890 Supplier Mettler Toledo Supplier No. 31052303
Add to Cart
Add to Cart
Detachable terminal with a 7 in. touchscreen for flexible placement options, increasing usability and convenience in 14 different language options. Compatible with Mettler Toledo NineFocus.
  • NineFocus pH/conductivity meter accessory
TRUSTED_SUSTAINABILITY
Specifications
Type Control Terminal
IP Rating IP52
Voltage 12 V
For Use With (Equipment) NineFocus pH Meter