Learn More
Medchemexpress LLC Ninerafaxstat (trihydrochloride) | 2311824-72-1 | 99.7% | 524.9 g/mol | C22H32Cl3N3O5 | 5 MG
SDP
Catalog No. 5000223143
Encompass_Preferred
Ninerafaxstat trihydrochloride is a research small-molecule salt investigated as a cardiac mitochondrial agent. It is provided for laboratory and preclinical studies and is supplied in solid and solution formats with accompanying analytical documentation.
  • Trihydrochloride salt form for improved stability and solubility.
  • Molecular weight 524.9 g/mol; CAS 2311824-72-1.
  • High purity (99.7%).
  • Available as solid vials (e.g., 5 mg) and solution (10 mM in DMSO).
  • Supplied with datasheet, COA, and SDS for traceability.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-23143 Supplier Medchemexpress LLC Supplier No. HY139577A5MG
Ninerafaxstat trihydrochloride is a research small-molecule salt investigated as a cardiac mitochondrial agent. It is provided for laboratory and preclinical studies and is supplied in solid and solution formats with accompanying analytical documentation.
  • Trihydrochloride salt form for improved stability and solubility.
  • Molecular weight 524.9 g/mol; CAS 2311824-72-1.
  • High purity (99.7%).
  • Available as solid vials (e.g., 5 mg) and solution (10 mM in DMSO).
  • Supplied with datasheet, COA, and SDS for traceability.
  • Intended for research use only; not for human therapeutic use.