Learn More
Medchemexpress LLC HY-107145A 5mg Medchemexpress, Ningetinib CAS:1394820-69-9 Purity:>98%
SDP
Catalog No. 501968367
Encompass_Preferred
Medchemexpress, HY-107145A 5mg Ningetinib CAS:1394820-69-9 Ningetinib is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and <1.0 nM for c-Met, VEGFR2 and Axl, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8367 Supplier Medchemexpress LLC Supplier No. HY107145A5MG
Medchemexpress, HY-107145A 5mg Ningetinib CAS:1394820-69-9 Ningetinib is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and <1.0 nM for c-Met, VEGFR2 and Axl, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.