Learn More
Medchemexpress LLC HY-107145 5mg Medchemexpress, Ningetinib Tosylate CAS:1394820-77-9 Purity:>98%
SDP
Catalog No. 501968366
Encompass_Preferred
Medchemexpress, HY-107145 5mg Ningetinib Tosylate CAS:1394820-77-9 Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and <1.0 nM for c-Met, VEGFR2 and Axl, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8366 Supplier Medchemexpress LLC Supplier No. HY1071455MG
Medchemexpress, HY-107145 5mg Ningetinib Tosylate CAS:1394820-77-9 Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and <1.0 nM for c-Met, VEGFR2 and Axl, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.