Learn More
Medchemexpress LLC Ninhydrin | 485-47-2 | 99.97% | 25 G
SDP
Catalog No. 5000414151
Encompass_Preferred
Ninhydrin is a chromogenic analytical probe used for the quantification of amino acids and proteins. It reacts with primary and secondary amines, producing a characteristic blue or purple reaction product: diketohydrindylidene-diketohydrindamine. Most amino acids, with the exception of proline, can be hydrolyzed and subsequently quantified colorimetrically following chromatographic separation. This product, appearing as a white to light yellow solid, is intended for research use only and has not been fully validated for medical applications.
  • Chromogenic analytical probe for the quantification of amino acids and proteins.
  • Reacts with primary and secondary amines.
  • Produces a blue or purple reaction product.
  • Suitable for the colorimetric quantification of most amino acids after chromatographic separation.
Catalog No. 50-004-14151 Supplier Medchemexpress LLC Supplier No. 25GHYD0908
Ninhydrin is a chromogenic analytical probe used for the quantification of amino acids and proteins. It reacts with primary and secondary amines, producing a characteristic blue or purple reaction product: diketohydrindylidene-diketohydrindamine. Most amino acids, with the exception of proline, can be hydrolyzed and subsequently quantified colorimetrically following chromatographic separation. This product, appearing as a white to light yellow solid, is intended for research use only and has not been fully validated for medical applications.
  • Chromogenic analytical probe for the quantification of amino acids and proteins.
  • Reacts with primary and secondary amines.
  • Produces a blue or purple reaction product.
  • Suitable for the colorimetric quantification of most amino acids after chromatographic separation.