Learn More
Medchemexpress LLC Ninhydrin | 485-47-2 | 99.99% | 50 G
SDP
Catalog No. 5000409730
Encompass_Preferred
Ninhydrin can be used as a chromogenic analytical probe for the quantification of amino acids and proteins. Most amino acids, except proline, can be hydrolyzed and react with ninhydrin. The amino acids are then quantified colorimetrically after separation by chromatography. Ninhydrin reacts with primary and secondary amines, producing a blue or purple reaction product: diketohydrindylidene-diketohydrindamine.
  • Chromogenic analytical probe
  • Used for the quantification of amino acids and proteins
  • Reacts with primary and secondary amines to produce a blue or purple product
Catalog No. 50-004-09730 Supplier Medchemexpress LLC Supplier No. HYD090850G
Ninhydrin can be used as a chromogenic analytical probe for the quantification of amino acids and proteins. Most amino acids, except proline, can be hydrolyzed and react with ninhydrin. The amino acids are then quantified colorimetrically after separation by chromatography. Ninhydrin reacts with primary and secondary amines, producing a blue or purple reaction product: diketohydrindylidene-diketohydrindamine.
  • Chromogenic analytical probe
  • Used for the quantification of amino acids and proteins
  • Reacts with primary and secondary amines to produce a blue or purple product