Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ninhydrin

Catalog No. N487625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N487625G 500 g
N4876100G 10 g
N487610G 25 g
N4876500G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. N487625G Supplier Sigma Aldrich Supplier No. N487625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ninhydrin

Specifications

CAS 485-47-2
Melting Point 250°C
Molecular Formula C9H6O4
Linear Formula C9H6O4
MDL Number MFCD00003791
Quantity 500 g
Synonym 1,2,3-Indantrione monohydrate; 2,2-Dihydroxy-1,3-indanedione; Trioxohydrindene monohydrate
Molecular Weight (g/mol) 178.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.