Learn More
Sigma Aldrich Fine Chemicals Biosciences Ninhydrin Reagent, 485-47-2, MFCD00003791, 100 mL
Catalog No. NC9040542
Encompass
Empirical Formula (Hill Notation): C9H6O4, Molecular Weight: 178.14, 2% Solution. Ninhydrin reagents can be used in thin layer chromatography (TLC). Ninhydrin reagents have been used to develop and validate a simple, selective, and precise densitometric method for analysis of α-aminocephalosporins, both in bulk drugs and in formulations. Ninhydrin reagents have also been used to observe differences in the reference ranges of several plasma and urine amino acids between Singaporean and Caucasian populations.
Catalog No. NC9040542 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N7285100ML
Empirical Formula (Hill Notation): C9H6O4, Molecular Weight: 178.14, 2% Solution. Ninhydrin reagents can be used in thin layer chromatography (TLC). Ninhydrin reagents have been used to develop and validate a simple, selective, and precise densitometric method for analysis of α-aminocephalosporins, both in bulk drugs and in formulations. Ninhydrin reagents have also been used to observe differences in the reference ranges of several plasma and urine amino acids between Singaporean and Caucasian populations.