Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin Reagent (According to Stahl for TLC derivatization), MilliporeSigma™ Supelco™

Catalog No. 111016745 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6745 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6745 Supplier MilliporeSigma Supelco Supplier No. 17975100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ninhydrin reagent is mostly used for the detection of amino acids. It basically gives a characteristic purple color on reaction with a-amino acids and other amino compounds, provided the pH of the reaction is 5.0 or higher. At an acid pH of 0.3, the reaction of ninhydrin with ornithinine and proline results in a red color product. Ninhydrin reagent can stay stable at room temperature in the absence of nitrogen atmosphere for upto 1 month in the ethylene glycol-sodium acetate solvent system.
TRUSTED_SUSTAINABILITY

Specifications

CAS 71-36-3
Density 0.823 g/mL (at 20°C)
Flash Point 34°C
Type Standard
Quantity 100 mL
Grade TLC Derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.