Learn More
Sigma Aldrich Fine Chemicals Biosciences Ninhydrin Spray Reagent, 100 mL
Catalog No. NC1529456
Encompass
Detects amino acids, amines and amino sugars.
Catalog No. NC1529456 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N1286100ML
Detects amino acids, amines and amino sugars.