Learn More
AnaSpec Ninhydrin Test Kit
Catalog No. 50848718
Encompass
Ninhydrin Test Kit[This test kit consists of three bottles of reagents. It is used to check if the coupling of a protected amino acid to the amino group of the resin is complete.] (1 each)
Catalog No. 50-848-718 Supplier AnaSpec Supplier No. AS25241
Ninhydrin Test Kit[This test kit consists of three bottles of reagents. It is used to check if the coupling of a protected amino acid to the amino group of the resin is complete.] (1 each)