Learn More
MCR Safety™ Ninja Ice Insulated Cut-Resistant Work Gloves, 7G Inside-15G Outside L
Catalog No. 19034378 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-378 Small
19-034-379 Medium
19-034-380 Large
19-034-377 X-Large
19-034-376 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-034-378 Supplier MCR Safety™ Supplier No. N9691S
Only null left
Add to Cart
Add to Cart
Ninja™ Ice insulated cut-resistant work gloves, 15-Gauge HyperMax™ Shell, 7-Gauge acrylic terry inner lining, HPT™ coated palm and fingertips.
  • 15-gauge cut-resistant HyperMax™ shell offers great cut protection
  • Acrylic terry lined insulated glove offers excellent cold protection while allowing air flow to help keep hands dry
  • Coating remains soft in temperatures as low as -4° F
  • Encapsulated air molecules provide a cushioning effect to minimize impact
  • The porous HPT coating with anti-wet treatment repels liquid for a firm, unmatched grip in wet or dry conditions
  • Treated with ActiFresh™ to help control bacteria and odor
Specifications
Construction String Knit
Coating Material PVC
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free No
Material Dyneema
Product Type Cut- and Abrasion-resistant Glove
Size Small
Gender Unisex
Glove Cuff Style Knit
Lining Lined
Puncture Rating ANSI/ISEA 105-16 Level 3
Thumb Style Wing