Learn More
MCR Safety™ Ninja Ice Insulated Work Gloves, 7G Inside-15G Outside
Catalog No. 19034383 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-383 Small
19-034-384 Medium
19-034-386 Large
19-034-382 X-Large
19-034-381 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-034-383 Supplier MCR Safety™ Supplier No. N9690S
Add to Cart
Add to Cart
Ninja™ Ice - insulated work gloves, 15-gauge black nylon with acrylic terry interior, HPT palm and fingertip coating.
  • Acrylic terry liner for insulation and warmth
  • Allows air to flow through which helps keep hands dry
  • Coating remains soft in temperatures as low as -4°F
  • Encapsulated air aolecules provide a cushioning effect to minimize impact
  • The porous HPT coating with Anti-Wet treatment repels liquid for an unprecedent firm grip in wet or dry conditions
  • Treated with ActiFresh™ to help control bacteria and odor
Specifications
ANSI Abrasion Rating ANSI 3
Chlorinated No
Construction String Knit
Coating Material PVC
Color Black
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Insulated Gloves
Size Small
Glove Cuff Style Knit
Lining Lined