Learn More
Medchemexpress LLC HY-50904 200mg Medchemexpress, Nintedanib CAS:656247-17-5 Purity:>98%
SDP
Catalog No. 502029352
Encompass_Preferred
Medchemexpress, HY-50904 200mg Nintedanib CAS:656247-17-5 Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9352 Supplier Medchemexpress LLC Supplier No. HY50904200MG
Medchemexpress, HY-50904 200mg Nintedanib CAS:656247-17-5 Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.