Learn More
Apexbio Technology LLC Nintedanib (BIBF 1120) 656247-17-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062512
Encompass_Preferred
Nintedanib (BIBF 1120 CAS 656247-17-5) is an orally bioavailable small-molecule tri-kinase inhibitor that targets vascular endothelial growth factor receptors (VEGFR1-3) fibroblast growth factor receptors (FGFR1-3) and platelet-derived growth factor receptors (PDGFR / ) By inhibiting these receptor pathways at nanomolar concentrations (IC50 20 100 nmol/L) Nintedanib disrupts angiogenesis signaling and demonstrates antiangiogenic activity It is extensively investigated in preclinical and clinical research for idiopathic pulmonary fibrosis (IPF) as well as oncology including non-small-cell lung carcinoma ovarian cancer colorectal cancer and hepatocellular carcinoma
Catalog No. 50-000-62512 Supplier Apexbio Technology LLC Supplier No. A82525.1
Nintedanib (BIBF 1120 CAS 656247-17-5) is an orally bioavailable small-molecule tri-kinase inhibitor that targets vascular endothelial growth factor receptors (VEGFR1-3) fibroblast growth factor receptors (FGFR1-3) and platelet-derived growth factor receptors (PDGFR / ) By inhibiting these receptor pathways at nanomolar concentrations (IC50 20 100 nmol/L) Nintedanib disrupts angiogenesis signaling and demonstrates antiangiogenic activity It is extensively investigated in preclinical and clinical research for idiopathic pulmonary fibrosis (IPF) as well as oncology including non-small-cell lung carcinoma ovarian cancer colorectal cancer and hepatocellular carcinoma