Learn More
Medchemexpress LLC Nintedanib-d8 | 1624587-87-6 | 99.9% | C31H25D8N5O4 | 5 MG
SDP
Catalog No. 5000358203
Encompass_Preferred
Nintedanib-d8 is a deuterium labeled Nintedanib, which is a potent triple angiokinase inhibitor targeting VEGFR1/2/3, FGFR1/2/3, and PDGFRα/β.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-58203 Supplier Medchemexpress LLC Supplier No. HY50904S25MG
Nintedanib-d8 is a deuterium labeled Nintedanib, which is a potent triple angiokinase inhibitor targeting VEGFR1/2/3, FGFR1/2/3, and PDGFRα/β.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.