Learn More
Medchemexpress LLC HY-11106 10mg Medchemexpress, Nintedanib esylate CAS:656247-18-6 Purity:>98%
SDP
Catalog No. 501968818
Encompass_Preferred
Medchemexpress, HY-11106 10mg Nintedanib esylate CAS:656247-18-6 Nintedanib esylate (BIBF 1120 esylate) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8818 Supplier Medchemexpress LLC Supplier No. HY1110610MG
Medchemexpress, HY-11106 10mg Nintedanib esylate CAS:656247-18-6 Nintedanib esylate (BIBF 1120 esylate) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.