Learn More
Strem, An Ascensus Company CAS# 12034-59-2. 50g. Niobium(IV) oxide, 99+%. MFCD00016242
SDP
Supplier:  Strem, An Ascensus Company 41411050G
Encompass
CAS# 12034-59-2. 50g. Niobium(IV) oxide, 99+%. MFCD00016242. Molecular Weight: 124.91. Molecular Formula: NbO2. Color/form: -200 mesh pwdr. Strem# 41-4110. http://www.strem.com/catalog/v/41-4110/
Catalog No. 50-901-11653