Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Niobium pentaoxide, 100g

Catalog No. 5000082544
Encompass_Preferred

Niobium pentaoxide 99 9% metals basisCas-1313-96-8

Catalog No. 50-000-82544 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. N108412100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Niobium pentaoxide 99 9% metals basisCas-1313-96-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.