Learn More
Strem, An Ascensus Company CAS# 7440-03-1. 25cm. Niobium rod (99.8%). MFCD00011126
SDP
Catalog No. 5090111640
Encompass
CAS# 7440-03-1. 25cm. Niobium rod (99.8%). MFCD00011126. Molecular Weight: 92.91. Molecular Formula: Nb. Color/form: 6.35 mm dia. (~2.7g/cm) Strem# 41-0070. http://www.strem.com/catalog/v/41-0070/
Catalog No. 50-901-11640 Supplier Strem, An Ascensus Company Supplier No. 41007025CM
CAS# 7440-03-1. 25cm. Niobium rod (99.8%). MFCD00011126. Molecular Weight: 92.91. Molecular Formula: Nb. Color/form: 6.35 mm dia. (~2.7g/cm) Strem# 41-0070. http://www.strem.com/catalog/v/41-0070/